Learn More
Cytiva Manifold Tray for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
GSA_VA
Catalog No. 45002617 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Ettan IPGphor 3 IEF Unit
Catalog No. For Use With (Equipment)
45-002-617 Ettan IPGphor 3 IEF Unit
1 options
Catalog No. 45-002-617 Supplier Cytiva Supplier No. 80649857
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Request a Quote
Manifold Tray for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
  • Uniform thermal characteristics
  • High heat conductivity
  • No wetted part
  • Sample equilibration in same tray
  • No. of gels: 1 to 12
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ettan IPGphor 3 IEF Unit
Type IPGphor Manifold Ceramic Tray
Material Ceramic
No. of Gels 1 to 12
Quantity Complete packsize