Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mannitol, 10% w/v, Reagents

Catalog No. 01334820
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-820 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-820 Supplier Reagents Holdings Llc Supplier No. CM0001001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mannitol, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 69-65-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym d-Mannitol
IUPAC Name water
SMILES O

Specifications

Boiling Range 100°C
Color Colorless
Boiling Point 100°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.0341
Percent Purity 10% w/v
Grade Reagent
Physical Form Liquid
Chemical Name or Material Mannitol, 10% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.