Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mannitol, Powder, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18604225 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-225 500 g Poly Bottle
18-604-224 125 g Poly Bottle
18-604-226 2.5 kg Poly Pail
18-604-227 12 kg Poly Pail
18-604-228 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-225 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MA165500GM
Only null left
Add to Cart
Add to Cart

MA165, 69-65-8, C6H14O6

Spectrum™ Chemical Mannitol, Powder, USP is a diuretic used to increase urine production.All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Melting Range 165-170°C
Packaging Poly Bottle
Quantity 500 g
Percent Purity 97 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.