Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mannose Phosphate Isomerase Antibody (OTI4G7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272584AF405
Description
Mannose Phosphate Isomerase Monoclonal specifically detects Mannose Phosphate Isomerase in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Mannose Phosphate Isomerase | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MPI | |
Full length recombinant protein of human Mannose Phosphate Isomerase (NP_002426) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4G7 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
CDG1B, EC 5.3.1, EC 5.3.1.8, FLJ39201, mannose phosphate isomerase, mannose-6- phosphate isomerase, mannose-6-phosphate isomerase, Phosphohexomutase, Phosphomannose isomerase, phosphomannose isomerase 1, PMI, PMI1phosphohexomutase | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
4351.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction