Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mannose Receptor Monoclonal Antibody (1C4), Bioss

Rabbit Monoclonal Antibody
Supplier: Bioss BSM52791R

This item is not returnable.
View return policy
Description
Mannose Receptor Monoclonal antibody specifically detects Mannose Receptor in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
Mannose Receptor | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 40% glycerol and 0.05% sodium azide | |
P22897 | |
Rabbit | |
Protein A | |
RUO | |
4360 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
1C4 | |
Unconjugated | |
Mrc1 | |
AW259686; bA541I19.1; CD206; CELE_Y39A3CR.4; CLEC13D; CLEC13DL; C-type lectin domain family 13 member D; C-type lectin domain family 13 member D-like; ddp-1; hMR; Human mannose receptor; macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1; mannose receptor, C type 1; mannose receptor, C type 1-like 1; Mitochondrial import inner membrane translocase subunit Tim8; MMR; MR; MRC1; MRC1L1; tim-8; Y39A3CR.4 | |
Full length protein (amino acids 1400-1456). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction