Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MANSC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989860
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9860 Supplier Bioss Supplier No. BS18656R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MANSC1 gene ontology (GO) annotation includes transmembrane transporter activity.

Specifications

Antigen MANSC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene MANSC1
Gene Accession No. Q9H8J5
Gene Alias 9130403P13Rik; LOH12CR3; Loss of heterozygosity 12 chromosomal region 3 protein; MANSC domain containing 1; MANSC domain-containing protein 1; MANSC1; UNQ316/PRO361
Gene Symbols MANSC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MANSC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54682
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.