Learn More
MilliporeSigma™ Supelco™ Manual Bubble Flowmeter, Capillary Version
Shop All MilliporeSigma Supelco Products
Click to view available options
Includes:
Flowmeter, Stand
Description
- A simple and precise way to measure flow rates of gases
- Allows the true split ratio to be determined by measuring actual flow rate
- The flow tube is graduated in 0.01 mL increments to accomodate low flows that are typical of capillary GC applications
- Two magnetic clamps hold the flow tube together so that the unit can be attached to any metal surface for easy storage
- A version that includes a sampling stand is offered for added portability
Specifications
Specifications
Type | Bubble Flowmeter |
For Use With (Application) | Measuring Gas Flow Rates |
Tube Size | 0.5 mL |
Includes | Flowmeter, Stand |