Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Manual Method Stain Wright Giemsa Stain and Matching Buffer

Catalog No. p-3605898
Click to view available options
Quantity:
2 x 250 mL
2 x 3800 mL (1 gal.)
This item is not returnable. View return policy
This item is not returnable. View return policy

Using flooding technique, this solution will stain all cellular elements without precipitate or artifactural changes. Manual Method Stain (Wright or Wright Giemsa). Available with and without matching buffer solution.

  • Solution uses flooding technique
  • Staining time: 5 to 7 minutes
  • Buffer is applied immediately after stain, reducing overall staining time
  • Convenient flip-top cap for 250 mL and 950 mL sizes
  • In the event of cold weather, allow solutions to return to room temperature

Specifications

Quantity 2 x 3800 mL (1 gal.)
Storage Requirements Room Temperature
For Use With (Application) For staining of cellular elements without precipitate or artifactural changes, in-vitro diagnostic use
Staining Time 5 to 7 min.
Packaging Bottles

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.