Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAO-A Mouse anti-Human, Clone: 5D1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275564
Description
MAO-A Monoclonal specifically detects MAO-A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MAO-A | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
amine oxidase [flavin-containing] A, EC 1.4.3, EC 1.4.3.4, MAO-A, monoamine oxidase A, Monoamine oxidase type A | |
Mouse | |
60 kDa | |
100 μL | |
4128.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
5D1 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide | |
MAOA | |
Recombinant protein within Human MAO-A aa 222-517/ 527. (SwissProt: P21397 Human) | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction