Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega MAO-Glo™ Monoamine Oxidase Assay System

Catalog No. PRV1402 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
200 Assays
Product Type:
Assay System
Monoamine Oxidase Assay Aystem
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV1402 1000 Assays Assay System
PRV1401 200 Assays Monoamine Oxidase Assay Aystem
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV1402 Supplier Promega Supplier No. V1402
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MAO-Glo™ Assay provides a homogeneous luminescent method for measuring monoamine oxidase activity from recombinant and native sources and for testing the effects of test compounds on MAO activity.

  • Sensitive Detection of Monoamine Oxidase (MAO) Activity
  • Luminescence-based method eliminates need for time-consuming analyses such as HPLC
  • Simple add and read protocol ideal for high-throughput screening
  • Less MAO enzyme required than typical HPLC or fluorometric methods

Specifications

For Use With (Application) Provides a homogeneous luminescent method for measuring MAO activity from recombinant and native sources
Content And Storage −30 to −10°C
Product Type Assay System
Quantity 1000 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.