Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAOA Monoclonal Antibody (5D1)
GREENER_CHOICE

Catalog No. PIMA534896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534896 Supplier Invitrogen™ Supplier No. MA534896
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: human liver tissue lysates, SH-SY-5Y.

This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAOA
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5D1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene MAOA
Gene Accession No. P21397
Gene Alias 1110061B18Rik; AA407771; Amine oxidase [flavin-containing] A; HGNC:6833; Mao; MAOA; MAO-A; monoamine oxidase A; monoamine oxidase type A; RP1-201D17__B.2
Gene Symbols MAOA
Host Species Mouse
Immunogen Recombinant protein within Human MAOA aa 222-517.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4128
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.