Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAOA Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518420
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome.
Specifications
MAOA | |
Polyclonal | |
Unconjugated | |
MAOA | |
1110061B18Rik; AA407771; Amine oxidase [flavin-containing] A; HGNC:6833; Mao; MAOA; MAO-A; monoamine oxidase A; monoamine oxidase type A; RP1-201D17__B.2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
4128 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P21397 | |
MAOA | |
Synthetic peptide sequence (DAPWEAQHADKWDK) corresponding to the internal amino acids of MAOA (aa 141-155). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction