Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAOB Recombinant Rabbit Monoclonal Antibody (9B6)
GREENER_CHOICE

Catalog No. PIMA538429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538429 Supplier Invitrogen™ Supplier No. MA538429
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody has been tested in direct-ELISA.

MAOB (Monoamine Oxidase B), also called MAO, BRAIN, AMINE OXIDASE (FLAVIN-CONTAINING) B, is a protein that in humans is encoded by the MAOB gene. MAOB is a member of the flavin monoamine oxidase family. And it is mapped on Xp11.3. MAOB catalyzes the oxidative deamination of biogenic and xenobiotic amines and plays an important role in the metabolism of neuroactive and vasoactive amines in the central nervous system and peripheral tissues. This protein preferentially degrades benzylamine and phenylethylamine. Like MAOA, it also degrades dopamine. MAO-B is involved in the breakdown of dopamine, a neurotransmitter implicated in reinforcing and motivating behaviors as well as movement. MAO-B inhibition is, therefore, associated with enhanced activity of dopamine, as well as with decreased production of hydrogen peroxide, a source of reactive oxygen species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAOB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 9B6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4
Gene MAOB
Gene Accession No. P27338
Gene Alias 6330414K01Rik; adrenalin oxidase; amine oxidase [flavin-containing] B; HGNC:6834; MAO b; MAO, brain; MAO, platelet; Maob; MAO-B; MGC26382; monoamine oxidase B; monoamine oxidase type B; monoamine oxidase-B; RP1-201D17__B.1; tyramine oxidase
Gene Symbols MAOB
Host Species Rabbit
Immunogen A synthesized peptide derived from human Monoamine Oxidase B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4129
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.