Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAOB Mouse anti-Human, Clone: 2B12F3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | MAOB |
|---|---|
| Clone | 2B12F3 |
| Concentration | 0.73 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6543
![]()
|
Proteintech Group Inc
661071IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The MAOB gene encodes a 520-amino acid protein with a molecular mass of 58.8 kDa and shows 70% amino acid identity to MAOA. MAOA and MAOB are present in the outer mitochondrial membrane in the central nervous system and peripheral tissues. A comparison of highly purified human placental MAOA and human liver MAOB revealed that the A form of the enzyme is larger by 2 kDa and only one potential N-glycosylation site exists in each protein, with each site in a different relevant position.Specifications
| MAOB | |
| 0.73 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| MAO B, MAOB, monoamine oxidase B, Monoamine oxidase type B | |
| MAOB | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2B12F3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P27338 | |
| 4129 | |
| MAOB Fusion Protein Ag17918 | |
| Primary | |
| -20°C | |
| MAOB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title