Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP-1 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277373
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7373 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7373 Supplier Leinco Technologies Inc Supplier No. 11009100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAP-1 Polyclonal antibody specifically detects MAP-1 in Human samples. It is validated for Western Blot

Specifications

Antigen MAP-1
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MOAP1
Gene Accession No. Q96BY2
Gene Alias AA987038; MAP1; Map-1; Moap1; Modulator of apoptosis 1; paraneoplastic antigen like 4; paraneoplastic antigen Ma4; paraneoplastic Ma antigen family member 4; PNMA4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa 337-351 of human MAP-1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64112
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.