Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP-2 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5622IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5622IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5622IMI Supplier MilliporeSigma Supplier No. AB5622I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-MAP-2 is a highly specific rabbit polyclonal antibody that targets microtubule-associated protein 2 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP-2
Applications Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Map2,Mtap2
Host Species Rabbit
Immunogen KLH-conjugated three peptides from N-and C-terminal regions of rat MAP-2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_037198
Test Specificity This polyclonal antibody detects MAP2 in Mouse and Rat tissues.
Target Species Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.