Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP 2a,b,c (Microtubule-Associated Protein) Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS250P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS250P 1 mL Concentrated 200μg/mL
MS250R7 7 mL Prediluted
MS250P0 100 μL Concentrated 200μg/mL
MS250P1 500 μL Concentrated 200μg/mL
MS250P1ABX 100 μL Concentrated 1mg/mL
MS250PABX 200 μL Concentrated 1mg/mL
MS250R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS250P Supplier Epredia Supplier No. MS250P

Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ MAP 2a,b,c (Microtubule-Associated Protein) Ab-3, Mouse Monoclonal Antibody.

Microtubule Associated Protein 2 exists in two high molecular weight forms (2a & 2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. Ab-3 is specific to phosphprylated form of MAP2a,b,c and shows no cross-reaction with other MAPs, tau, and tubulin. Ab-3 reacts with dendrites and cell bodies of neurons. It is useful in studies of neuron structure in normal and malignant brain tissue and in Alzheimer's.

Host Species: Mouse

Clone: AP18

Isotype: IgG1

Species Reactivity: Human, Cow, Rat, Xenopus, Quail. Others not known.

Epitope: aa 1-151

Immunogen: Bovine brain microtubule associated protein 2

Molecular Weight: ∽280kDa doublet and a 70kDa singlet

Positive Control: IMR-5 or T98G human glioblastoma cells or brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Specifications

Antigen MAP 2a,b,c (Microtubule-Associated Protein) Ab-3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone AP18
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Bovine brain microtubule associated protein 2
Quantity 1 mL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Bovine, Human, Quail, Rat, Xenopus species
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.