Learn More
MAP 2a,b,c (Microtubule-Associated Protein) Ab-3 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: AP18
Isotype: IgG1
Species Reactivity: Human, Cow, Rat, Xenopus, Quail. Others not known.
Epitope: aa 1-151
Immunogen: Bovine brain microtubule associated protein 2
Molecular Weight: ∽280kDa doublet and a 70kDa singlet
Positive Control: IMR-5 or T98G human glioblastoma cells or brain
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Specifications
| Antigen | MAP 2a,b,c (Microtubule-Associated Protein) Ab-3 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | AP18 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Bovine brain microtubule associated protein 2 |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.