Learn More
Epredia™ Lab Vision™ MAP 2a,b (Microtubule-Associated Protein) Ab-2, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ MAP 2a,b (Microtubule-Associated Protein) Ab-2, Mouse Monoclonal Antibody.
$579.00
Specifications
Antigen | MAP 2a,b (Microtubule-Associated Protein) Ab-2 |
---|---|
Clone | AP20 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS249S1
|
Epredia™
MS249S1 |
Concentrated | 500 μL |
Each for $579.00
|
|
||||
MS249S0
|
Epredia™
MS249S0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS249S
|
Epredia™
MS249S |
Concentrated | 1 mL | N/A | N/A | N/A | |||
MS249R7
|
Epredia™
MS249R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
Microtubule Associated Protein 2 exists in two high molecular weight forms (2a & 2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. Ab-2 is highly specific to MAP2a,b and shows no cross-reaction with MAP1, MAP5, tubulin, or tau. Ab-2 reacts with dendrites and cell bodies of neurons but not with the neuronal processes. It does not react with the low molecular weight (70kDa) form (2c) of MAP2. It is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's.Host Species: Mouse
Clone: AP20
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: aa 997-1332
Immunogen: Bovine brain microtubule associated protein 2
Molecular Weight: ∽280kDa Doublet
Positive Control: Human glioblastoma T98G cells or brain
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
MAP 2a,b (Microtubule-Associated Protein) Ab-2 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RUO | |
Human | |
IgG1 |
AP20 | |
Monoclonal | |
Mouse | |
Neuroscience | |
Bovine brain microtubule associated protein 2 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.