Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562625 Supplier Invitrogen™ Supplier No. PA562625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HEHPPMLVNP PHIQNIGATN QTCQTEQNRI NTIRQLPLLN ALLVELSLLY DQPVTSPAHI HPHLAWLYRT EDKKSPESSA KSTCRSEAKK DKRSVG Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 57%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP10
Gene Accession No. Q9P2G4
Gene Alias 4933403G14Rik; AI835009; KIAA1383; Map10; microtubule associated protein 10; microtubule regulator 120 KDa; microtubule regulator of 120 KDa; Microtubule-associated protein 10; mKIAA1383; MTR120
Gene Symbols MAP10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAP10. Recombinant protein control fragment (Product #RP-99828).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54627
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.