Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Map17 Antibody, Novus Biologicals™
SDP

Catalog No. p-5082638 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400151SS 0.025 mL
NB400151 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400151SS Supplier Novus Biologicals Supplier No. NB400151SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Map17 Polyclonal specifically detects Map17 in Human, Mouse, Rat samples. It is validated for Western Blot, ICC/IF (Negative).

Specifications

Antigen Map17
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ICC/IF (Negative)
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. Q9CQH0
Gene Alias 17 kDa membrane-associated protein, DD96, MAP17epithelial protein up-regulated in carcinoma, membrane associated protein 17, membrane-associated protein 17, PDZK1 interacting protein 1, PDZK1-interacting protein 1, Protein DD96, RP1-18D14.5, SPAP
Gene Symbols PDZK1IP1
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of mouse Map17. [UniProt# Q9CQH0]
Molecular Weight of Antigen 14 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10158
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.