Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APSA6820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APSA6820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APSA6820 Supplier Invitrogen™ Supplier No. PA316820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is not recommended for ICC/IF applications.

The small PDZK1-associated protein (SPAP; also known as MAP17) is an endogenous regulator of cellular PDZK1 levels by regulating PDZK1 turnover.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP17
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene PDZK1IP1
Gene Accession No. Q13113, Q923S2, Q9CQH0
Gene Alias 0610007F13Rik; 17 kDa membrane-associated protein; 2700030M23Rik; AU046042; DD96; epithelial protein up-regulated in carcinoma, membrane associated protein 17; MAP17; membrane-associated protein 17; membrane-associated protein 17 (Map17); PDZK1 interacting protein 1; PDZK1-interacting protein 1; PDZK1IP1; Protein DD96; SPAP
Gene Symbols PDZK1IP1
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of mouse MAP17.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10158, 67182, 81916
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.