Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP1A Antibody (HM-1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001371
Description
MAP1A Monoclonal specifically detects MAP1A in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
MAP1A | |
Monoclonal | |
Unconjugated | |
EC 2.7.2.8, EC 6.3.4.5, FLJ77111, MAP-1A, MAP1LMTAP1A, microtubule-associated protein 1A, Proliferation-related protein p80 | |
Mouse | |
Unpurified | |
RUO | |
4130 | |
Mouse, Rat | |
Ascites |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
HM-1 | |
Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
MAP1A | |
Rat brain MAPs preparation. | |
0.1 mL | |
Primary | |
does not stain tubulin or other microtubule associated proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction