Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1A/MAP1B LC3 A/B Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501985005
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5005 Supplier Bioss Supplier No. BS2912R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Structural protein involved in the filamentous cross-bridging between microtubules and other skeletal elements.

Specifications

Antigen MAP1A/MAP1B LC3 A/B
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAP1A
Gene Accession No. P34926, P78559, Q9QYR6
Gene Alias 6330416M19Rik; AI853608; MAP1 light chain LC2; Map1a; MAP-1A; MAP1A heavy chain; MAP1L; microtubule associated protein 1A; microtubule-associated protein 1 A; Microtubule-associated protein 1A; modifier of tubby hearing 1; moth1; Mtap1; Mtap-1; Mtap1a; Proliferation-related protein p80
Gene Symbols MAP1A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAP1A/MAP1B LC3 B .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17754, 25152, 4130
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.