Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP1A Monoclonal Antibody (HM-1)
GREENER_CHOICE

Catalog No. PIMA532988
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532988 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532988 Supplier Invitrogen™ Supplier No. MA532988
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute by adding 1 mL of PBS buffer to yield a concentration of 100 μg/mL. Positive Control - IHC: Rat Brain Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Structural protein involved in the filamentous cross-bridging between microtubules and other skeletal elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1A
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HM-1
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide
Gene MAP1A
Gene Accession No. P34926, Q9QYR6
Gene Alias 6330416M19Rik; AI853608; MAP1 light chain LC2; MAP1A; MAP-1A; MAP1A heavy chain; MAP1L; microtubule associated protein 1A; microtubule-associated protein 1 A; microtubule-associated protein 1A; modifier of tubby hearing 1; moth1; Mtap1; Mtap-1; Mtap1a; Proliferation-related protein p80
Gene Symbols MAP1A
Host Species Mouse
Immunogen Rat brain microtubule-associated proteins (MAPs).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17754, 25152
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.