Learn More
Invitrogen™ MAP1A Polyclonal Antibody

Description
Immunogen sequence: VQEWQETSPT REEPAGEQKE LAPAWEDTSP EQDNRYWRGR EDVALEQDTY WRELSCERKV WFPHELDGQG ARPHYTEERE STFLDEGPDD EQEVPLREHA TRSPWASDFK DFQESSPQK Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 73%.
Specifications
Specifications
| Antigen | MAP1A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MAP1A |
| Gene Accession No. | P78559 |
| Gene Alias | 6330416M19Rik; AI853608; MAP1 light chain LC2; MAP1A; MAP-1A; MAP1A heavy chain; MAP1L; microtubule associated protein 1A; microtubule-associated protein 1 A; microtubule-associated protein 1A; modifier of tubby hearing 1; moth1; Mtap1; Mtap-1; Mtap1a; Proliferation-related protein p80 |
| Gene Symbols | MAP1A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.