Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566845 Supplier Invitrogen™ Supplier No. PA566845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGEREEEGRAE APDKSSHSSK VPEASKSHAT TEPEQTEPEQ REPTPYPDER SFQYADIYEQ MMLTGLGPAC PTREPPLGAA GDWPPCLSTK EAAAGRNTSA EKELSSPISP K Highest antigen sequence identity to the following orthologs - mouse 83%, rat 83%.

Structural protein involved in the filamentous cross-bridging between microtubules and other skeletal elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP1A
Gene Accession No. P78559
Gene Alias 6330416M19Rik; AI853608; MAP1 light chain LC2; MAP1A; MAP-1A; MAP1A heavy chain; MAP1L; microtubule associated protein 1A; microtubule-associated protein 1 A; microtubule-associated protein 1A; modifier of tubby hearing 1; moth1; Mtap1; Mtap-1; Mtap1a; Proliferation-related protein p80
Gene Symbols MAP1A
Host Species Rabbit
Immunogen Recombinant Human MAP1A. Recombinant protein control fragment (Product #RP-107499).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4130
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.