Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP1B Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10068256FR
Description
MAP1B Polyclonal specifically detects MAP1B in Human, Mouse samples. It is validated for Western Blot.Specifications
MAP1B | |
Polyclonal | |
Western Blot | |
DKFZp686E1099, DKFZp686F1345, FLJ38954, FUTSCH, MAP-1B, MAP5, microtubule-associated protein 1B | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAP1B | |
The immunogen recognized by this antibody maps to a region between residue 900 and 950 of human microtubule-associated protein 1B using the numbering given in entry NP_005900.2 (GeneID 4131). | |
100 μL | |
Cytoskeleton Markers, MAP Kinase Signaling, Neuronal Cell Markers | |
4131 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction