Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS248R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS248R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS248R1 Supplier Epredia™ Supplier No. MS248R1

Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1, Mouse Monoclonal Antibody.

Microtubule-associated protein 1B (MAP1B, also known as MAP5, MAP1.2, MAP1(x), or MAP1X) is different from MAP1A (also known as MAP1 or MAP1.1). Reportedly, MAP1B/MAP5 is an early MAP; it is present at high levels in embryonic and newborn rat brain and declines several-fold upon brain maturation. In several cellular situations, MAP1B/MAP5 is the first neuronal MAP to appear, and it is found in neurites from their very first emergence from the cell body. Expression of MAP1B/MAP5 is induced by nerve growth factor.

Host Species: Mouse

Clone: 3G5

Isotype: IgG1

Species Reactivity: Human, Cow and Rat. Does not react with chicken. Others not known.

Epitope: Not determined

Immunogen: Purified bovine brain MAPs

Molecular Weight: 320kDa doublet

Positive Control: Brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3G5
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Purified bovine brain MAPs
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Bovine, Human, Rat
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.