Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530584
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530584 Supplier Invitrogen™ Supplier No. PA530584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, P4.5 mouse brain(24ug), Neuro-2a(15ug). Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1B
Applications Western Blot
Classification Polyclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene MAP1B
Gene Accession No. P14873, P46821
Gene Alias A230055D22; AI843217; claustrin; DKFZp686E1099; DKFZp686F1345; FLJ38954; futsch; I79_010564; LC1; MAP1 light chain LC1; MAP1(X); MAP1.2; MAP1B; MAP-1B; MAP1B heavy chain; MAP5; microtubule associated protein 1B; microtubule associated protein 1B; microtubule-associated protein 1B; microtubule-associated protein 1B; Microtubule-associated protein 1B (MAP 1B) (MAP1.2) (MAP1(X)); microtubule-associated protein 5; Mtap1b; Mtap5; Mtap-5; Neuraxin; PPP1R102; protein phosphatase 1, regulatory subunit 102
Gene Symbols MAP1B
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MAP1B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17755, 4131
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.