Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582798 Supplier Invitrogen™ Supplier No. PA582798
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVVEEHCASP EDKTLEVVSP SQSVTGSAGH TPYYQSPTDE KSSHLPTEVI EKPPAVPVSF EFSDAKDENE RASVSPMDEP VPDSESPIEK VLSPLRSPPL IGSESAYESF LSADDKASGR GAESPFEEKS GKQGSPDQVS PVSEMTST Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 85%.

The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP1B
Gene Accession No. P46821
Gene Alias A230055D22; AI843217; claustrin; DKFZp686E1099; DKFZp686F1345; FLJ38954; futsch; I79_010564; LC1; MAP1 light chain LC1; MAP1(X); MAP1.2; MAP1B; MAP-1B; MAP1B heavy chain; MAP5; microtubule associated protein 1B; microtubule associated protein 1B; microtubule-associated protein 1B; microtubule-associated protein 1B; Microtubule-associated protein 1B (MAP 1B) (MAP1.2) (MAP1(X)); microtubule-associated protein 5; Mtap1b; Mtap5; Mtap-5; Neuraxin; PPP1R102; protein phosphatase 1, regulatory subunit 102
Gene Symbols MAP1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAP1B. Recombinant protein control fragment (Product #RP-102411).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4131
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.