Learn More
MAP1B Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301447AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that belongs to the microtubule-associated protein family. The proteins of this family are thought to be involved in microtubule assembly, which is an essential step in neurogenesis. The product of this gene is a precursor polypeptide that presumably undergoes proteolytic processing to generate the final MAP1B heavy chain and LC1 light chain. Gene knockout studies of the mouse microtubule-associated protein 1B gene suggested an important role in development and function of the nervous system.Specifications
| MAP1B | |
| Polyclonal | |
| Unconjugated | |
| MAP1B | |
| DKFZp686E1099, DKFZp686F1345, FLJ38954, FUTSCH, MAP5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4131 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| P46821 | |
| MAP1B | |
| Between 2050 and 2100 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.