Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1LC3B Rabbit anti-Human, Mouse, Rat, Clone:RM293, Abnova™

Catalog No. 89333263 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-263 Supplier Abnova Corporation Supplier No. MAB16916
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against human MAP1LC3B.

The product of this gene is a subunit of neuronal microtubule-associated MAP1A and MAP1B proteins, which are involved in microtubule assembly and important for neurogenesis. Studies on the rat homolog implicate a role for this gene in autophagy, a process that involves the bulk degradation of cytoplasmic component. [provided by RefSeq]

Specifications

Antigen MAP1LC3B
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RM293
Conjugate Unconjugated
Description microtubule-associated protein 1 light chain 3 beta
Dilution Immunocytochemistry (1:100-200)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-200)
Western Blot (1:100-200)
The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene MAP1LC3B
Gene Alias LC3B, MAP1A/1BLC3
Host Species Rabbit
Immunogen A peptide corresponding to N-terminus of human MAP1LC3B.
Quantity 100 μL
Gene ID (Entrez) 81631
Test Specificity This antibody reacts to human LC3B (Autophagy-related protein LC3 B) (Microtubule-associated proteins 1A/1B light chain 3B). It may also react to mouse or rat LC3B, as predicted by immunogen homology.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.