Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP1LC3B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144237
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144237 Supplier Invitrogen™ Supplier No. PA5144237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAP1LC3B2 Polyclonal Antibody for ICC/IF, ELISA

Ubiquitin-like modifier involved in formation of autophagosomal vacuoles (autophagosomes). Plays a role in mitophagy which contributes to regulate mitochondrial quantity and quality by eliminating the mitochondria to a basal level to fulfill cellular energy requirements and preventing excess ROS production. In response to cellular stress and upon mitochondria fission, binds C-18 ceramides and anchors autophagolysosomes to outer mitochondrial membranes to eliminate damaged mitochondria. While LC3s are involved in elongation of the phagophore membrane, the GABARAP/GATE-16 subfamily is essential for a later stage in autophagosome maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1LC3B2
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene MAP1LC3B2
Gene Accession No. A6NCE7
Gene Alias ATG8G; MAP1LC3B2; Microtubule associated protein 1 light chain 3 beta 2; Microtubule associated proteins 1A/1B light chain 3B like protein; Microtubule-associated proteins 1A/1B light chain 3 beta 2; Microtubule-associated proteins 1A/1B light chain 3B-like; MP3B2_HUMAN
Gene Symbols MAP1LC3B2
Host Species Rabbit
Immunogen Recombinant Human Microtubule-associated proteins 1A/1B light chain 3 beta 2 protein (1-47AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643246
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.