Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP1LC3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111700 Supplier Invitrogen™ Supplier No. PA5111700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TKFLVPQELT MTQFLSIIRS RMVLRATEAF Highest antigen sequence indentity to the following orthologs: Rat - 50%, Mouse - 50%.

MAP1LC3C, also named as LC3C, belongs to the MAP1 LC3 family. It is the human homolog of yeast Atg8 and is involved in the formation of autophagosomal vacuoles, called autophagosomes. LC3C is produced by the proteolytic cleavage after the conserved C-terminal Gly residue. The antibody is specific to the prepeptide of LC3C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1LC3C
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP1LC3C
Gene Accession No. Q9BXW4
Gene Alias ATG8J; Autoph; autophagy-related protein LC3 C; Autophagy-related ubiquitin-like modifier LC3 C; LC3C; LC3-like protein 2; MAP1 light chain 3-like protein 2; MAP1 light chain 3-like protein 3; MAP1A/MAP1B LC3 C; MAP1A/MAP1B light chain 3 C; MAP1LC3C; microtubule associated protein 1 light chain 3 gamma; microtubule-associated protein 1 light chain 3 gamma; Microtubule-associated proteins 1A/1B light chain 3C
Gene Symbols MAP1LC3C
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human MAP1LC3C. Recombinant protein control fragment (Product #RP-107891).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440738
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.