Learn More
Invitrogen™ MAP1LC3C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121185
Description
Positive test controls include: NCI-H460, OVCAR3, HepG2, 293T, Rat lung. The target is usually found in the following locations: Cytoplasm, Cytoplasmic vesicle, Endomembrane system, Lipid-anchor, autophagosome, autophagosome membrane, cytoskeleton. Immunogen sequence: MPPPQKIPSV RPFKQRKSLA IRQEEVAGIR AKFPNKIPVV VERYPRETFL PPLDKTKFLV PQELTMTQFL SIIRSRMVLR ATEAFYLLVN NKSLVSMSAT MAEIYRDYKD EDGFVYMTYA SQETFGCLES AAPRDGSSLE DRPCNPL.
MAP1LC3C, also named as LC3C, belongs to the MAP1 LC3 family. It is the human homolog of yeast Atg8 and is involved in the formation of autophagosomal vacuoles, called autophagosomes. LC3C is produced by the proteolytic cleavage after the conserved C-terminal Gly residue. The antibody is specific to the prepeptide of LC3C.
Specifications
MAP1LC3C | |
Polyclonal | |
Unconjugated | |
MAP1LC3C | |
ATG8J; Autoph; autophagy-related protein LC3 C; Autophagy-related ubiquitin-like modifier LC3 C; LC3C; LC3-like protein 2; MAP1 light chain 3-like protein 2; MAP1 light chain 3-like protein 3; MAP1A/MAP1B LC3 C; MAP1A/MAP1B light chain 3 C; MAP1LC3C; microtubule associated protein 1 light chain 3 gamma; microtubule-associated protein 1 light chain 3 gamma; Microtubule-associated proteins 1A/1B light chain 3C | |
Rabbit | |
Affinity chromatography | |
RUO | |
440738 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.92 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9BXW4 | |
MAP1LC3C | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human MAP1LC3C (NP_001004343.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.