Learn More
MAP1LC3C Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against partial synthetic protein of human MAP1LC3C.
Supplier: Abnova Corporation PAB29560
Description
Autophagy is a highly regulated bulk degradation process that plays an important role in cellular maintenance and development. MAP1LC3C is 1 of 3 orthologs of the yeast autophagosome protein Atg8 (He et al., 2003 [PubMed 12740394]).[supplied by OMIM]
Specifications
MAP1LC3C | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial synthetic protein of human MAP1LC3C. | |
In PBS (0.09% sodium azide) | |
MAP1LC3C | |
A synthetic peptide (conjugated with KLH) corresponding to amino acids 1-30 at N-terminus of human MAP1LC3C. | |
400 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
LC3C | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
440738 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Ig |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.