Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP1S Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP147282C
Description
MAP1S Polyclonal antibody specifically detects MAP1S in Human samples. It is validated for Western BlotSpecifications
MAP1S | |
Polyclonal | |
Western Blot | |
BPY2 interacting protein 1, BPY2IP1VCY2IP-1, C19orf5, chromosome 19 open reading frame 5, FLJ10669, MAP8MAP-1S, MGC133087, microtubule-associated protein 1S, Microtubule-associated protein 8, Variable charge Y chromosome 2-interacting protein 1, VCY2 interacting protein 1, VCY2-interacting protein 1, VCY2IP1BPY2-interacting protein 1 | |
The immunogen for this product maps to a region between residue 525 and 575 of human MAP1S using the numbering given in entry NP_060644.4 (GeneID 55201). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55201 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction