Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP1S Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562272 Supplier Invitrogen™ Supplier No. PA562272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GFGVPRHDPL PDPLKVPPPL PDPSSICMVD PEMLPPKTAR QTENVSRTRK PLARPNSRAA APKATPVAAA KTKGLAGGDR ASRPLSAR Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 62%.

Microtubule-associated protein that mediates aggregation of mitochondria resulting in cell death and genomic destruction (MAGD). Plays a role in anchoring the microtubule organizing center to the centrosomes. Binds to DNA. Plays a role in apoptosis. Involved in the formation of microtubule bundles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1S
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP1S
Gene Accession No. Q66K74
Gene Alias BPY2 interacting protein 1; BPY2-interacting protein 1; BPY2IP1; C19orf5; MAP1S; MAP-1S; MAP1S heavy chain; MAP1S light chain; MAP8; microtubule associated protein 1S; microtubule-associated protein 1S; Microtubule-associated protein 8; variable charge Y chromosome 2-interacting protein 1; VCY2 interacting protein 1; VCY2-interacting protein 1; VCY2IP1; VCY2IP-1
Gene Symbols MAP1S
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAP1S. Recombinant protein control fragment (Product #RP-101273).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55201
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.