Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP1S Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100607 Supplier Invitrogen™ Supplier No. PA5100607
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAP1S.

Microtubule-associated protein that mediates aggregation of mitochondria resulting in cell death and genomic destruction (MAGD). Plays a role in anchoring the microtubule organizing center to the centrosomes. Binds to DNA. Plays a role in apoptosis. Involved in the formation of microtubule bundles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1S
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAP1S
Gene Accession No. P0C5W1, Q66K74, Q8C052
Gene Alias BPY2 interacting protein 1; BPY2-interacting protein 1; BPY2IP1; C19orf5; MAP1S; MAP-1S; MAP1S heavy chain; MAP1S light chain; MAP8; microtubule associated protein 1S; microtubule-associated protein 1S; Microtubule-associated protein 8; variable charge Y chromosome 2-interacting protein 1; VCY2 interacting protein 1; VCY2-interacting protein 1; VCY2IP1; VCY2IP-1
Gene Symbols MAP1S
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAP1S(Accession Q66K74), corresponding to amino acid residues S273-D323.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270058, 290640, 55201
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.