Learn More
MAP1S Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302815A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Microtubule-associated protein that mediates aggregation of mitochondria resulting in cell death and genomic destruction (MAGD). Plays a role in anchoring the microtubule-organizing center to the centrosomes. Binds to DNA. Plays a role in apoptosis. Involved in the formation of microtubule bundles.Specifications
MAP1S | |
Polyclonal | |
Unconjugated | |
MAP1S | |
BPY2IP1, C19orf5, MAP8, VCY2IP-1, VCY2IP1, BPY2 interacting protein 1, BPY2-interacting protein 1, MAP-1S, VCY2 interacting protein 1, VCY2-interacting protein 1, microtubule-associated protein 8, variable charge Y chromosome 2-interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55201 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q66K74 | |
MAP1S | |
Between 475 and 525 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.