Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Antibody (4H5), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 2 publications

$243.50 - $512.00

Specifications

Antigen MAP2
Clone 4H5
Concentration 1 mg/mL
Dilution Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:300, Immunohistochemistry Free-Floating 1:2000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP225156SS
SDP
View Documents
Novus Biologicals
NBP225156SS
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NBP225156
SDP
View Documents
Novus Biologicals
NBP225156
0.1 mL
Each for $512.00
Only null left
Add to Cart
 
Description

Description

MAP2 Monoclonal specifically detects MAP2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating.
Specifications

Specifications

MAP2
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat, Bovine
P11137
4133
MAP2 Antibody (4H5) was developed against full length purified bovine protein, epitope mapped to projection domain of human sequence, between amino acids 631 and 1056.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
199 kDa
4H5
Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:300, Immunohistochemistry Free-Floating 1:2000
Monoclonal
Mouse
Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
50% PBS, 50% glycerol with 5mM Sodium Azide
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
MAP2
IgG1
Affinity Purified
MAP2 Antibody (4H5) will be reactive to isoforms 1(MAP2B) and isoform 3 (MAP2A).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.