Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP2 Antibody (5H11), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192711AF350
Description
MAP2 Monoclonal specifically detects MAP2 in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.Specifications
MAP2 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAP2 | |
MAP2 Antibody (5H11) was developed against full length purified bovine protein, epitope mapped to projection domain of human sequence, amino acids 1057-1588 using MAP2 Projection Domain 3. | |
0.1 mL | |
Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
Note that since the epitope for this antibody in within the projection domain found only in MAP2A and MAP2B, and so the antibody does not bind to the lower molecular weight MAP2C and MAP2D isoforms which lack this region. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
5H11 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating | |
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction