Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Antibody (5H11), Novus Biologicals™
SDP

Catalog No. NBP192711SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192711SS 0.025 mL
NBP192711 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP192711SS Supplier Novus Biologicals Supplier No. NBP192711SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

MAP2 Monoclonal specifically detects MAP2 in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.

Specifications

Antigen MAP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone 5H11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Accession No. P11137
Gene Alias DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
Gene Symbols MAP2
Host Species Mouse
Immunogen MAP2 Antibody (5H11) was developed against full length purified bovine protein, epitope mapped to projection domain of human sequence, amino acids 1057-1588 using MAP2 Projection Domain 3.
Molecular Weight of Antigen 199 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4133
Test Specificity Note that since the epitope for this antibody in within the projection domain found only in MAP2A and MAP2B, and so the antibody does not bind to the lower molecular weight MAP2C and MAP2D isoforms which lack this region.
Target Species Human, Mouse, Rat, Bovine, Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.