Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP2 Antibody (HM-2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12011267
Description
MAP2 Monoclonal specifically detects MAP2 in Human, Mouse, Rat, Avian, Bovine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.Specifications
MAP2 | |
Monoclonal | |
2.0 mg/mL | |
Western Blot 1-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Microarray | |
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
Mouse | |
280 kDa | |
0.05 mL | |
Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
4133 | |
Human, Mouse, Rat, Avian, Bovine, Chicken | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HM-2 | |
Unconjugated | |
P15146 | |
MAP2 | |
This MAP2 antibody was developed against rat brain microtubule associated proteins (MAPs). | |
Affinity Purified | |
RUO | |
Primary | |
Reacts with all known forms of MAP2, namely MAP2a, MAP2b and MAP2c. Does not cross-react with other MAPs or tubulin. By immunohistochemical staining of formalin-fixed, paraffin-embedded brain tissue, the antibody shows selective labeling of dendritic trees throughout the brain. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction