Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Mouse anti-Human, Clone: M13, Abnova™

Catalog No. 89035516 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-516 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-516 Supplier Abnova Corporation Supplier No. MAB14118

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human MAP2.

  • High specificity
  • High reactivity

Specifications

Antigen MAP2
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M13
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human MAP2.
Dilution ELISA (0.1-0.5 ug/mL) Flow Cytometry (3-5 ug x 106 cells) Immunocytochemistry (1-2 ug/mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (5-10 ug/mL) Immunoprecipitation (2-5 ug) Western Blot (0.5-2 ug/mL) The optimal
Formulation In PBS, pH 7.4 (0.09% sodium azide).
Gene Accession No. P11137
Gene Alias DKFZp686I2148/MAP2A/MAP2B/MAP2C
Gene Symbols MAP2
Host Species Mouse
Immunogen MAPs.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4133
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.