Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP2 Mouse, DyLight 350, Clone: HM-2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12011267UV0.1ML
Description
MAP2 Monoclonal specifically detects MAP2 in Human, Mouse, Rat, Avian, Bovine, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MAP2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAP2 | |
Rat brain microtubule associated proteins (MAPs). | |
0.1 mL | |
Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuroscience, Phospho Specific, Stem Cell Markers | |
4133 | |
Human, Mouse, Rat, Avian, Bovine, Chicken | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HM-2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, microtubule-associated protein 2 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Monoclonal Anti-MAP2 does not react with tubulin of other MAPs or tubulin. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction