Learn More
MAP2K1 Monoclonal Antibody (OTI3C1), TrueMAB™, OriGene
SDP
Mouse Monoclonal Antibody
Supplier:  OriGene TA506069
Encompass_Preferred
 View more versions of this product
Catalog No. 50-167-6425
Only null left
Add to Cart
Description
Description
MEK1 (mitogen activated protein kinase kinase, ERK kinase 1, microtubule-associated protein 2 kinase) is a dual specificity kinase and catalyses both a threonine and a tyrosine residue on MAP kinases ERK1 and ERK2. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in MEK1 can lead to cardiofaciocutaneous syndrome.
Specifications
Specifications
MAP2K1
Monoclonal
Unconjugated
MAP2K1
CFC3, MAPKK1, MEK1, MKK1, PRKMK1
Mouse
Affinity Chromatography
RUO
26395, 478347, 5604
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3C1
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P31938, Q02750
MAP2K1
Full length human recombit protein of human MAP2K1 produced in HEK293T cell.
100 μL
Primary
Canine, Human, Mouse
Antibody
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.