Learn More
MAP2K1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1F9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506016

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MEK1 (mitogen activated protein kinase kinase, ERK kinase 1, microtubule-associated protein 2 kinase) is a dual specificity kinase and catalyses both a threonine and a tyrosine residue on MAP kinases ERK1 and ERK2. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in MEK1 can lead to cardiofaciocutaneous syndrome.Specifications
MAP2K1 | |
Monoclonal | |
Unconjugated | |
MAP2K1 | |
CFC3, MAPKK1, MEK1, MKK1, PRKMK1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
170851, 26395, 478347, 5604 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1F9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P31938, Q01986, Q02750 | |
Map2k1 | |
Full length human recombit protein of human MAP2K1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.