Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2K1, Rabbit anti-Human, Clone: D3, Abnova™

Catalog No. 89335781 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-781 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-781 Supplier Abnova Corporation Supplier No. MAB18947
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen MAP2K1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone D3
Conjugate Unconjugated
Dilution Flow Cytometry Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide).
Gene mitogen-activated protein kinase kinase 1
Gene Alias MAPKK1/MEK1/MKK1/PRKMK1
Gene Symbols MAP2K1
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding S221 of human MAP2K1/MAP2K2.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5604/5605
Target Species Human, Mouse
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.